(R)-2-([2,2'-Bipyridin]-6-yl)-4-isopropyl-4,5-dihydrooxazole 97%
Supplier: Ambeed
|
Warning
|
A1351976-100MG
A1351976-1G
A1351976-250MG
76658-446EA
148.53
USD
76658-446
76658-448
76658-450
(R)-2-([2,2'-Bipyridin]-6-yl)-4-isopropyl-4,5-dihydrooxazole 97%
(R)-2-([2,2'-Bipyridin]-6-yl)-4-isopropyl-4,5-dihydrooxazole
Precaución: for Research Use Only
Formula:
C₁₆H₁₇N₃O MW: 267.3257 g/mol |
CAS Number:
2757082-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...