(S)-2-([2,2'-Bipyridin]-6-yl)-4-phenyl-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1351974-100MG A1351974-1G A1351974-250MG
76658-440EA 144 USD
76658-440 76658-442 76658-444
(S)-2-([2,2'-Bipyridin]-6-yl)-4-phenyl-4,5-dihydrooxazole ≥97%, ee 99%
(S)-2-([2,2'-Bipyridin]-6-yl)-4-phenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₉H₁₅N₃O
MW: 301.3419 g/mol
CAS Number: 915314-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR