(4R,4'R)-4,4'-Diisobutyl-4,4',5,5'-tetrahydro-2,2'-bioxazole 97%
Supplier: Ambeed
|
Warning
|
A1351972-1G
A1351972-250MG
76658-436EA
944.21
USD
76658-436
76658-438
(4R,4'R)-4,4'-Diisobutyl-4,4',5,5'-tetrahydro-2,2'-bioxazole 97%
(4R,4'R)-4,4'-Diisobutyl-4,4',5,5'-tetrahydro-2,2'-bioxazole
Precaución: for Research Use Only
Formula:
C₁₄H₂₄N₂O₂ MW: 252.3526 g/mol |
CAS Number:
2270178-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...