2,6-Bis((R)-4-isobutyl-4,5-dihydrooxazol-2-yl)pyridine ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
A1351969-100MG
A1351969-1G
A1351969-250MG
76658-430EA
59
USD
76658-430
76658-432
76658-434
2,6-Bis((R)-4-isobutyl-4,5-dihydrooxazol-2-yl)pyridine ≥97%, ee 99%
2,6-Bis((R)-4-isobutyl-4,5-dihydrooxazol-2-yl)pyridine
Precaución: for Research Use Only
Formula:
C₁₉H₂₇N₃O₂ |
CAS Number:
1658490-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...