(S)-4-Isopropyl-2-(6-phenylpyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1351957-100MG A1351957-1G A1351957-250MG
76658-390EA 83 USD
76658-390 76658-392 76658-394
(S)-4-Isopropyl-2-(6-phenylpyridin-2-yl)-4,5-dihydrooxazole ≥97%, ee 99%
(S)-4-Isopropyl-2-(6-phenylpyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O
CAS Number: 1509929-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR