(S)-4-(tert-Butyl)-2-(pyrimidin-2-yl)-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1351950-100MG A1351950-1G A1351950-250MG A1351950-50MG
76658-356EA 79.57 USD
76658-356 76658-358 76658-360 76658-362
(S)-4-(tert-Butyl)-2-(pyrimidin-2-yl)-4,5-dihydrooxazole 97%
(S)-4-(tert-Butyl)-2-(pyrimidin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₃O
MW: 205.2563 g/mol
CAS Number: 2070868-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR