4,4'-(2,2-Diphenylethene-1,1-diyl)bis(bromobenzene) 97%

Supplier: Ambeed
Warning

A1351861-1G A1351861-250MG A1351861-5G
76658-348EA 35 USD
76658-348 76658-350 76658-352
4,4'-(2,2-Diphenylethene-1,1-diyl)bis(bromobenzene) 97%
4,4'-(2,2-Diphenylethene-1,1-diyl)bis(bromobenzene)

Precaución: for Research Use Only

Formula: C₂₆H₁₈Br₂
MDL Number: MFCD32876950
CAS Number: 859315-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR