4-(1,2,2-Triphenylvinyl)benzonitrile 97%

Supplier: Ambeed
Warning

A1351856-1G A1351856-250MG A1351856-5G
76658-330EA 25 USD
76658-330 76658-332 76658-334
4-(1,2,2-Triphenylvinyl)benzonitrile 97%
4-(1,2,2-Triphenylvinyl)benzonitrile

Precaución: for Research Use Only

Formula: C₂₇H₁₉N
CAS Number: 1446909-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR