4',4''',4''''',4'''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1'-biphenyl]-4-ol)) 97%

Supplier: Ambeed
Warning

A1351849-1G A1351849-250MG A1351849-5G
76658-312EA 92.3 USD
76658-312 76658-314 76658-316
4',4''',4''''',4'''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1'-biphenyl]-4-ol)) 97%
4',4''',4''''',4'''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1'-biphenyl]-4-ol))

Precaución: for Research Use Only

Formula: C₅₀H₃₆O₄
MW: 700.8185 g/mol
CAS Number: 1712454-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR