4',4''',4''''',4'''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1'-biphenyl]-4-carbonitrile)) 97%

Supplier: Ambeed
Warning

A1351848-1G A1351848-250MG
76658-308EA 73.21 USD
76658-308 76658-310
4',4''',4''''',4'''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1'-biphenyl]-4-carbonitrile)) 97%
4',4''',4''''',4'''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1'-biphenyl]-4-carbonitrile))

Precaución: for Research Use Only

Formula: C₅₄H₃₂N₄
MW: 736.8587 g/mol
CAS Number: 608129-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR