3,3'-Dihydroxy-[1,1'-biphenyl]-4,4'-dicarbaldehyde 97%

Supplier: Ambeed
Warning

A1351789-100MG A1351789-1G A1351789-250MG
76658-288EA 48.81 USD
76658-288 76658-290 76658-292
3,3'-Dihydroxy-[1,1'-biphenyl]-4,4'-dicarbaldehyde 97%
3,3'-Dihydroxy-[1,1'-biphenyl]-4,4'-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₄
MW: 242.2268 g/mol
CAS Number: 14969-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR