4,4'-(3,8-Bis(4-aminophenyl)pyrene-1,6-diyl)dibenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A1351775-100MG
A1351775-25MG
A1351775-50MG
76658-278EA
220
USD
76658-278
76658-280
76658-282
4,4'-(3,8-Bis(4-aminophenyl)pyrene-1,6-diyl)dibenzaldehyde 97%
4,4'-(3,8-Bis(4-aminophenyl)pyrene-1,6-diyl)dibenzaldehyde
Precaución: for Research Use Only
Formula:
C₄₂H₂₈N₂O₂ |
CAS Number:
2375652-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...