4,4'-(3,8-Bis((4-aminophenyl)ethynyl)pyrene-1,6-diyl)dibenzaldehyde 97%

Supplier: Ambeed
Warning

A1351774-100MG A1351774-1G A1351774-250MG
76658-272EA 119 USD
76658-272 76658-274 76658-276
4,4'-(3,8-Bis((4-aminophenyl)ethynyl)pyrene-1,6-diyl)dibenzaldehyde 97%
4,4'-(3,8-Bis((4-aminophenyl)ethynyl)pyrene-1,6-diyl)dibenzaldehyde

Precaución: for Research Use Only

Formula: C₄₆H₂₈N₂O₂
CAS Number: 2375652-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR