4,4',4'',4'''-([2,2'-Bi(1,3-dithiolylidene)]-4,4',5,5'-tetrayl)tetraaniline 97%
Supplier: Ambeed
|
Warning
|
A1351768-100MG
A1351768-1G
A1351768-250MG
76658-266EA
190.97
USD
76658-266
76658-268
76658-270
4,4',4'',4'''-([2,2'-Bi(1,3-dithiolylidene)]-4,4',5,5'-tetrayl)tetraaniline 97%
4,4',4'',4'''-([2,2'-Bi(1,3-dithiolylidene)]-4,4',5,5'-tetrayl)tetraaniline
Precaución: for Research Use Only
Formula:
C₃₀H₂₄N₄S₄ MW: 568.8000 g/mol |
CAS Number:
2413296-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...