2,3,5,6-Tetraaminobenzoquinone 97%

Supplier: Ambeed
Warning

A1351766-100MG A1351766-1G A1351766-250MG A1351766-5G
76658-260EA 20 USD
76658-260 76658-262 76658-264 77867-428
2,3,5,6-Tetraaminobenzoquinone 97%
2,3,5,6-Tetraaminobenzoquinone

Precaución: for Research Use Only

Formula: C₆H₈N₄O₂
MDL Number: MFCD18449057
CAS Number: 1128-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR