5',5''-Bis(4-aminophenyl)-[1,1':3',1'':3'',1'''-quaterphenyl]-4,4'''-diamine 97%

Supplier: Ambeed
Warning

A1351765-100MG A1351765-1G A1351765-250MG A1351765-5G
76658-254EA 62 USD
76658-254 76658-256 76658-258 77867-427
5',5''-Bis(4-aminophenyl)-[1,1':3',1'':3'',1'''-quaterphenyl]-4,4'''-diamine 97%
5',5''-Bis(4-aminophenyl)-[1,1':3',1'':3'',1'''-quaterphenyl]-4,4'''-diamine

Precaución: for Research Use Only

Formula: C₃₆H₃₀N₄
CAS Number: 2411406-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR