4,4'-(9,10-Anthracenediyl)bis[benzenamine] 97%

Supplier: Ambeed
Warning

A1351763-100MG A1351763-1G A1351763-250MG A1351763-5G
76658-246EA 23 USD
76658-246 76658-248 76658-250 76658-252
4,4'-(9,10-Anthracenediyl)bis[benzenamine] 97%
4,4'-(9,10-Anthracenediyl)bis[benzenamine]

Precaución: for Research Use Only

Formula: C₂₆H₂₀N₂
MDL Number: MFCD00356298
CAS Number: 106704-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR