(2S,2'S)-2,2'-(1,3,8,10-Tetraoxoanthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-2,9(1H,3H,8H,10H)-diyl)dipropanoic acid 97%

Supplier: Ambeed
Warning

A1351758-100MG A1351758-1G A1351758-250MG A1351758-5G
76658-238EA 47.75 USD
76658-238 76658-240 76658-242 76658-244
(2S,2'S)-2,2'-(1,3,8,10-Tetraoxoanthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-2,9(1H,3H,8H,10H)-diyl)dipropanoic acid 97%
(2S,2'S)-2,2'-(1,3,8,10-Tetraoxoanthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-2,9(1H,3H,8H,10H)-diyl)dipropanoic acid

Precaución: for Research Use Only

Formula: C₃₀H₁₈N₂O₈
MW: 534.4725 g/mol
CAS Number: 925685-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR