3,3'-(Pyridine-3,5-diyl)dibenzoic acid 97%

Supplier: Ambeed
Warning

A1351754-100MG A1351754-1G A1351754-250MG A1351754-5G
76658-230EA 8.49 USD
76658-230 76658-232 76658-234 76658-236
3,3'-(Pyridine-3,5-diyl)dibenzoic acid 97%
3,3'-(Pyridine-3,5-diyl)dibenzoic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₃NO₄
MW: 319.3108 g/mol
CAS Number: 1429436-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR