N1,N2-Dimethyl-N1,N2-bis(pyridin-2-ylmethyl)ethane-1,2-diamine 97%

Supplier: Ambeed
Warning

A1351737-5G A1351737-100MG
76658-218EA 475.29 USD
76658-218 77621-644
N1,N2-Dimethyl-N1,N2-bis(pyridin-2-ylmethyl)ethane-1,2-diamine 97%
N1,N2-Dimethyl-N1,N2-bis(pyridin-2-ylmethyl)ethane-1,2-diamine

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₄
MW: 270.3727 g/mol
MDL Number: MFCD29039185
CAS Number: 136768-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR