2-Amino-6-(propylamino)-5,6-dihydrobenzo[d]thiazol-7(4H)-one hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A1351624-100MG
A1351624-250MG
76658-214EA
137.92
USD
76658-214
76658-216
2-Amino-6-(propylamino)-5,6-dihydrobenzo[d]thiazol-7(4H)-one hydrochloride 98%
2-Amino-6-(propylamino)-5,6-dihydrobenzo[d]thiazol-7(4H)-one hydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₁₆ClN₃OS MW: 261.7715 g/mol |
MDL Number:
MFCD24386471 CAS Number: 2245708-68-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...