2-Methyl-2-phenoxy-1-phenylpropan-1-ol 97%

Supplier: Ambeed
Warning

A1351407-100MG A1351407-1G A1351407-250MG
76658-198EA 73.21 USD
76658-198 76658-200 76658-202
2-Methyl-2-phenoxy-1-phenylpropan-1-ol 97%
2-Methyl-2-phenoxy-1-phenylpropan-1-ol

Precaución: for Research Use Only

Formula: C₁₆H₁₈O₂
MW: 242.3129 g/mol
CAS Number: 29509-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR