2,3-Bis(4-(diphenylamino)phenyl)benzo[b]thiophene 1,1-dioxide 97%

Supplier: Ambeed
Warning

A1351275-1G A1351275-25G A1351275-5G
76658-192EA 26 USD
76658-192 76658-194 76658-196
2,3-Bis(4-(diphenylamino)phenyl)benzo[b]thiophene 1,1-dioxide 97%
2,3-Bis(4-(diphenylamino)phenyl)benzo[b]thiophene 1,1-dioxide

Precaución: for Research Use Only

Formula: C₄₄H₃₂N₂O₂S
CAS Number: 2091868-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR