Tetrakis(4-ethynylphenyl)methane 98% (by GC)

Supplier: Ambeed
Warning

A999603-100MG A999603-1G A999603-250MG
76657-810EA 133.68 USD
76657-810 76657-812 76657-814
Tetrakis(4-ethynylphenyl)methane 98% (by GC)
Tetrakis(4-ethynylphenyl)methane

Precaución: for Research Use Only

Formula: C₃₃H₂₀
MW: 416.52 g/mol
MDL Number: MFCD24873189
CAS Number: 177991-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR