Pentadecanal 98%

Supplier: Ambeed
Warning

Synonyms: Pentadecanal, Pentadecylaldehyde

A999513-100MG A999513-1G A999513-250MG A999513-5G
76657-804EA 42.44 USD
76657-804 76657-806 76657-808 77599-460
Pentadecanal 98%
Pentadecanal

Precaución: for Research Use Only

Formula: C₁₅H₃₀O
MW: 226.4 g/mol
MDL Number: MFCD12911660
CAS Number: 2765-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR