2-(2-Nitrovinyl)furan 98%

Supplier: Ambeed
Danger

Synonyms: 1-(Fur-2-yl)-2-nitroethene, 1-(2-Furyl)-2-nitroethylene, 2-(2-Nitroethenyl)furan, 1-(Fur-2-yl)-2-nitroethylene

A999401-1G A999401-25G A999401-5G
76657-798EA 12.74 USD
76657-798 76657-800 76657-802
2-(2-Nitrovinyl)furan 98%
2-(2-Nitrovinyl)furan

Precaución: for Research Use Only

Formula: C₆H₅NO₃
MW: 139.109 g/mol
MDL Number: MFCD00030600
CAS Number: 699-18-3
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR