Meso-Tetra (3-pyridyl) porphine 97%

Supplier: Ambeed
Warning

A999097-100MG A999097-1G A999097-250MG A999097-5G
76657-784EA 15 USD
76657-784 76657-786 76657-788 76657-790
Meso-Tetra (3-pyridyl) porphine 97%
Meso-Tetra (3-pyridyl) porphine

Precaución: for Research Use Only

Formula: C₄₀H₂₆N₈
MW: 618.6881 g/mol
MDL Number: MFCD00600955
CAS Number: 40882-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR