1-Propyl-3-methylimidazolium bis((trifluoromethyl)sulfonyl)imide 99%

Supplier: Ambeed
Danger

A997753-1G A997753-25G
76657-778EA 6 USD
76657-778 76657-780
1-Propyl-3-methylimidazolium bis((trifluoromethyl)sulfonyl)imide 99%
1-Propyl-3-methylimidazolium bis((trifluoromethyl)sulfonyl)imide

Precaución: for Research Use Only

Formula: C₉H₁₃F₆N₃O₄S₂
MW: 405.34 g/mol
MDL Number: MFCD11111918
CAS Number: 216299-72-8
UN: 2922
ADR: 8,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR