1-Ethyl-3-methylimidazoliumtrifluoroacetate 99%

Supplier: Ambeed
Warning

Synonyms: 1-ethyl-3-methylimidazolium trifluoroacetate

A997523-1G A997523-25G
76657-774EA 9.55 USD
76657-774 76657-776
1-Ethyl-3-methylimidazoliumtrifluoroacetate 99%
1-Ethyl-3-methylimidazolium trifluoroacetate

Precaución: for Research Use Only

Formula: C₈H₁₁F₃N₂O₂
MW: 224.1804 g/mol
MDL Number: MFCD08458492
CAS Number: 174899-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR