4-Methylbenzamidoxime 97%

Supplier: Ambeed
Warning

A996467-25G A996467-250MG A996467-1G A996467-5G
76657-754EA 254 USD
76657-754 77784-757 77784-756 77784-758
4-Methylbenzamidoxime 97%
4-Methylbenzamidoxime

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MDL Number: MFCD00019952
CAS Number: 19227-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR