2-Bromopyrazolo[1,5-a]pyrimidin-5(4H)-one 95%

Supplier: Ambeed
Warning

A995782-100MG A995782-1G A995782-250MG
76657-722EA 130.5 USD
76657-722 76657-724 76657-726
2-Bromopyrazolo[1,5-a]pyrimidin-5(4H)-one 95%
2-Bromopyrazolo[1,5-a]pyrimidin-5(4H)-one

Precaución: for Research Use Only

Formula: C₆H₄BrN₃O
MW: 214.0195 g/mol
MDL Number: MFCD31699410
CAS Number: 2357108-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR