Benzyl 1-acetylcyclopropane-1-carboxylate 97%

Supplier: Ambeed
Warning

A995771-100MG A995771-1G A995771-250MG
76657-716EA 42 USD
76657-716 76657-718 76657-720
Benzyl 1-acetylcyclopropane-1-carboxylate 97%
Benzyl 1-acetylcyclopropane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₄O₃
CAS Number: 664364-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR