4-Methoxyphenyl acetate 95%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxyphenylacetate

A995329-250MG A995329-100MG
76657-700EA 63 USD
76657-700 77623-436
4-Methoxyphenyl acetate 95%
4-Methoxyphenyl acetate

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MDL Number: MFCD00048211
CAS Number: 1200-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR