RU-521 98%

Supplier: Ambeed
Warning

Synonyms: 3-[1-(6,7-dichloro-1H-benzimidazol-2-yl)-5-hydroxy-3-methyl-pyrazol-4-yl]-3H-isobenzofuran-1-one, RU.521, 3-(1-(6,7-Dichloro-1H-benzo[d]imidazol-2-yl)-5-hydroxy-3-methyl-1H-pyrazol-4-yl)isobenzofuran-1(3H)-one

A995309-10MG A995309-1MG A995309-25MG A995309-5MG A995309-50MG
76657-692EA 136 USD
76657-692 76657-694 76657-696 76657-698 77749-829
RU-521 98%
RU-521

Precaución: for Research Use Only

Formula: C₁₉H₁₂Cl₂N₄O₃
MW: 415.23 g/mol
MDL Number: MFCD32173679
CAS Number: 2262452-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR