2-(Methanesulfonyloxy)ethyl methanesulfonate 95%

Supplier: Ambeed
Warning

Synonyms: Ethylene dimethanesulphonate

A995299-100MG A995299-1G A995299-250MG
76657-686EA 9.55 USD
76657-686 76657-688 76657-690
2-(Methanesulfonyloxy)ethyl methanesulfonate 95%
2-(Methanesulfonyloxy)ethyl methanesulfonate

Precaución: for Research Use Only

Formula: C₄H₁₀O₆S₂
MW: 218.2486 g/mol
MDL Number: MFCD00025060
CAS Number: 4672-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR