(3aR,5R,6S,6aR)-5-(((tert-Butyldiphenylsilyl)oxy)methyl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol 95%

Supplier: Ambeed
Warning

A994984-100MG A994984-1G A994984-250MG A994984-5G
76657-674EA 18.04 USD
76657-674 76657-676 76657-678 76657-680
(3aR,5R,6S,6aR)-5-(((tert-Butyldiphenylsilyl)oxy)methyl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol 95%
(3aR,5R,6S,6aR)-5-(((tert-Butyldiphenylsilyl)oxy)methyl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol

Precaución: for Research Use Only

Formula: C₂₄H₃₂O₅Si
MW: 428.5934 g/mol
MDL Number: MFCD22666287
CAS Number: 114861-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR