4',4''',4''''',4'''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1'-biphenyl]-4-carbaldehydE)) 97%

Supplier: Ambeed
Warning

A994799-100MG A994799-250MG A994799-5G A994799-1G
76657-666EA 26.53 USD
76657-666 76657-668 76657-670 77595-610
4',4''',4''''',4'''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1'-biphenyl]-4-carbaldehydE)) 97%
4',4''',4''''',4'''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1'-biphenyl]-4-carbaldehydE))

Precaución: for Research Use Only

Formula: C₅₄H₃₆O₄
MW: 748.8613 g/mol
MDL Number: MFCD31700762
CAS Number: 1624970-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR