Methyl 3-oxo-1,2,3,4-tetrahydroisoquinoline-7-carboxylate 95%

Supplier: Ambeed
Warning

A994742-100MG A994742-1G A994742-250MG
76657-660EA 96.55 USD
76657-660 76657-662 76657-664
Methyl 3-oxo-1,2,3,4-tetrahydroisoquinoline-7-carboxylate 95%
Methyl 3-oxo-1,2,3,4-tetrahydroisoquinoline-7-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
MW: 205.2099 g/mol
MDL Number: MFCD27952400
CAS Number: 1823924-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR