N-(3-(4-Fluorophenylsulfonamido)-4-methoxyphenyl)-[1,1'-biphenyl]-4-carboxamide 95%

Supplier: Ambeed
Warning

Synonyms: SN-001

A994632-100MG A994632-1G
76657-656EA 84.88 USD
76657-656 76657-658
N-(3-(4-Fluorophenylsulfonamido)-4-methoxyphenyl)-[1,1'-biphenyl]-4-carboxamide 95%
N-(3-(4-Fluorophenylsulfonamido)-4-methoxyphenyl)-[1,1'-biphenyl]-4-carboxamide

Precaución: for Research Use Only

Formula: C₂₆H₂₁FN₂O₄S
MW: 476.5194 g/mol
MDL Number: MFCD04617604
CAS Number: 727699-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR