[Ir{dFCF3ppy}2(bpy)]PF6 97%

Supplier: Ambeed
Warning

A993547-100MG A993547-1G A993547-250MG A993547-50MG
76657-622EA 44 USD
76657-622 76657-624 76657-626 76657-628
[Ir{dFCF3ppy}2(bpy)]PF6 97%
[Ir{dFCF3ppy}2(bpy)]PF6

Precaución: for Research Use Only

Formula: C₃₄H₁₈F₁₆IrN₄P
MDL Number: MFCD28987331
CAS Number: 1092775-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR