1-(2-(Trifluoromethyl)pyridin-4-yl)ethanamine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A993345-100MG
A993345-250MG
76657-618EA
213.25
USD
76657-618
76657-620
1-(2-(Trifluoromethyl)pyridin-4-yl)ethanamine hydrochloride 95%
1-(2-(Trifluoromethyl)pyridin-4-yl)ethanamine hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₀ClF₃N₂ MW: 226.6266 g/mol |
MDL Number:
MFCD28656550 CAS Number: 1951440-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...