(R)-2-((tert-butoxycarbonyl)amino)-2-(naphthalen-2-yl)acetic acid 95%

Supplier: Ambeed
Warning

A993148-100MG A993148-1G A993148-250MG
76657-612EA 71.09 USD
76657-612 76657-614 76657-616
(R)-2-((tert-butoxycarbonyl)amino)-2-(naphthalen-2-yl)acetic acid 95%
(R)-2-((tert-butoxycarbonyl)amino)-2-(naphthalen-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₉NO₄
MW: 301.3371 g/mol
MDL Number: MFCD07371919
CAS Number: 93779-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR