4,4-Dimethylcyclohexane-1-carbonitrile 95%

Supplier: Ambeed
Warning

A992981-100MG A992981-1G A992981-250MG
76657-606EA 186.72 USD
76657-606 76657-608 76657-610
4,4-Dimethylcyclohexane-1-carbonitrile 95%
4,4-Dimethylcyclohexane-1-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₁₅N
MW: 137.2221 g/mol
MDL Number: MFCD20319755
CAS Number: 25186-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR