2-Bromo-n-(tert-butyl)acetamide 95%

Supplier: Ambeed
Warning

A992972-100MG A992972-1G A992972-250MG A992972-5G A992972-10G
76657-598EA 13 USD
76657-598 76657-600 76657-602 76657-604 77784-700
2-Bromo-n-(tert-butyl)acetamide 95%
2-Bromo-n-(tert-butyl)acetamide

Precaución: for Research Use Only

Formula: C₆H₁₂BrNO
MDL Number: MFCD04113978
CAS Number: 57120-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR