Ethyl 7,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate 95%

Supplier: Ambeed
Warning

A992893-100MG A992893-1G A992893-250MG A992893-50MG
76657-586EA 53 USD
76657-586 76657-588 76657-590 76657-592
Ethyl 7,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate 95%
Ethyl 7,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₉F₂NO₃
MW: 253.2016 g/mol
MDL Number: MFCD12674971
CAS Number: 185011-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR