1-(10H-phenothiazin-2-yl)propan-1-one 95%

Supplier: Ambeed
Warning

A992751-100G A992751-25G A992751-5G
76657-564EA 157.02 USD
76657-564 76657-566 76657-568
1-(10H-phenothiazin-2-yl)propan-1-one 95%
1-(10H-phenothiazin-2-yl)propan-1-one

Precaución: for Research Use Only

Formula: C₁₅H₁₃NOS
MW: 255.3348 g/mol
MDL Number: MFCD02929294
CAS Number: 92-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR