2-(2-(2-(2-Bromoethoxy)ethoxy)ethoxy)ethanol 97%

Supplier: Ambeed
Warning

Synonyms: Br-PEG4-OH

A992732-100MG A992732-1G A992732-250MG A992732-5G A992732-25G
76657-556EA 30.77 USD
76657-556 76657-558 76657-560 76657-562 77623-418
2-(2-(2-(2-Bromoethoxy)ethoxy)ethoxy)ethanol 97%
2-(2-(2-(2-Bromoethoxy)ethoxy)ethoxy)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₇BrO₄
MW: 257.1222 g/mol
MDL Number: MFCD18205852
CAS Number: 85141-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR