1-Phenylhexane-1,3,5-trione 95%

Supplier: Ambeed
Warning

A992684-100MG A992684-1G A992684-250MG
76657-550EA 137.92 USD
76657-550 76657-552 76657-554
1-Phenylhexane-1,3,5-trione 95%
1-Phenylhexane-1,3,5-trione

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₃
MW: 204.2219 g/mol
MDL Number: MFCD00839524
CAS Number: 1469-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR