(Difluoromethyl)triphenylphosphonium bromide 97%

Supplier: Ambeed
Warning

A992596-100MG A992596-1G A992596-250MG A992596-5G
76657-534EA 90 USD
76657-534 76657-536 76657-538 76657-540
(Difluoromethyl)triphenylphosphonium bromide 97%
(Difluoromethyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₁₉H₁₆BrF₂P
MW: 393.2000 g/mol
MDL Number: MFCD33021715
CAS Number: 58310-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR