5-Bromo-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 98%

Supplier: Ambeed
Warning

A992553-100MG A992553-1G A992553-250MG A992553-25G A992553-5G
76657-524EA 6 USD
76657-524 76657-526 76657-528 76657-530 76657-532
5-Bromo-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 98%
5-Bromo-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₃H₉BrN₂O₄
MDL Number: MFCD30188073
CAS Number: 26166-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR